Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145303.2 | NP_001138775.2 | 2 | 363 | ||
NM_144686.3 | NP_653287.2 | 2 | 345 | |||
XM_011526486.2 | XP_011524788.1 | 2 | 328 | |||
GenBank mRNA | AK314973.1 | - | 2 | 266 | ||
AY236492.1 | - | 2 | 362 | |||
AY263166.1 | - | 2 | 242 | |||
BC025323.1 | - | 2 | 252 | 2959382 | ||
BQ082417.1 | - | 2 | 255 | |||
DQ656026.2 | - | 2 | 191 |