Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282607.1 | NP_001269536.1 | 12 | 1850 | ||
NM_144967.3 | NP_659404.2 | 12 | 2086 | |||
XM_006724729.1 | XP_006724792.1 | 11 | 1557 | |||
XM_011531280.1 | XP_011529582.1 | 11 | 1574 | |||
GenBank mRNA | AK054620.1 | - | 9 | 1446 | ||
AK128203.1 | - | 12 | 2086 | |||
AK294274.1 | - | 12 | 1809 | |||
AK296748.1 | - | 12 | 1828 | |||
BC063790.1 | - | 12 | 1685 | 6196091 |