Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHAL6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144071.1 | NP_001137543.1 | 6 | 905 | ||
NM_144972.4 | NP_659409.2 | 5 | 1498 | |||
XM_005252805.4 | XP_005252862.2 | 6 | 787 | |||
XM_011519922.2 | XP_011518224.1 | 7 | 843 | |||
XM_011519923.2 | XP_011518225.1 | 7 | 930 | |||
XM_011519924.2 | XP_011518226.1 | 7 | 787 | |||
XM_011519925.1 | XP_011518227.1 | 7 | 769 | |||
GenBank mRNA | AK131523.1 | - | 5 | 1441 | ||
AY581313.1 | - | 6 | 905 | |||
BC014340.1 | - | 6 | 909 | 3935569 |