Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CES5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143685.1 | NP_001137157.1 | 3 | 484 | ||
NM_001190158.1 | NP_001177087.1 | 4 | 568 | |||
NM_145024.2 | NP_659461.1 | 3 | 484 | |||
GenBank mRNA | AK056109.1 | - | 3 | 484 | ||
AK090997.1 | - | 3 | 410 | |||
AK294334.1 | - | 4 | 568 | |||
AY907669.1 | - | 3 | 411 | |||
BC039073.1 | - | 3 | 365 | |||
BC069501.1 | - | 3 | 455 | 7262427 | ||
BC069548.1 | - | 3 | 454 | 7262439 | ||
BC117126.1 | - | 3 | 457 | 40125677 | ||
BC143692.1 | - | 3 | 457 | 9052206 | ||
EU794678.1 | - | 3 | 484 |