Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CNTRL SIRNA Assays ›
Gene Symbol:
CNTRL
Gene Name:
centriolin
Gene Aliases:
CEP1, CEP110, FAN, bA165P4.1
Chromosome Location:
Chr.9: 121074778 - 121177610 on Build GRCh38
UniGene ID:
Hs.653263
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
3779
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CNTRL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_007018.4
NP_008949.4
25
4371
XM_005251679.3
XP_005251736.1
27
4643
XM_006716940.2
XP_006717003.1
27
4643
XM_006716942.1
XP_006717005.1
16
2872
XM_006716944.3
XP_006717007.1
27
4643
XM_011518166.1
XP_011516468.1
27
4643
XM_011518167.1
XP_011516469.1
26
4484
XM_011518168.1
XP_011516470.1
27
4568
XM_011518169.2
XP_011516471.1
26
4518
XM_011518170.1
XP_011516472.1
26
4466
XM_011518172.2
XP_011516474.1
27
4643
XM_011518173.1
XP_011516475.1
15
2718
XM_017014223.1
XP_016869712.1
27
4643
XM_017014224.1
XP_016869713.1
27
4643
XM_017014225.1
XP_016869714.1
25
4329
XM_017014226.1
XP_016869715.1
16
2872
XM_017014227.1
XP_016869716.1
26
4661
XM_017014228.1
XP_016869717.1
26
4517
XM_017014229.1
XP_016869718.1
27
4643
XM_017014230.1
XP_016869719.1
26
4434
GenBank mRNA
AF083322.1
-
2
819
AF513978.1
-
25
4340
AK074079.1
-
16
2744
AK093457.1
-
8
1141
AK297330.1
-
691
AY651261.1
-
18
3043
BC002932.2
-
16
2826
BC009287.2
-
12
2090
BC137284.1
-
25
4364
9021281
BC137286.1
-
25
4363
9021283
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
peptidyl-tyrosine phosphorylation
cell division
protein tyrosine kinase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0