Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMILIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007046.3 | NP_008977.1 | 2 | 726 | ||
GenBank mRNA | AF088916.1 | - | 2 | 227 | ||
AK075517.1 | - | 2 | 651 | |||
AK307539.1 | - | 2 | 655 | |||
AL050138.1 | - | 2 | 157 | |||
AL552691.3 | - | 2 | 732 | |||
BC007530.1 | - | 2 | 398 | |||
BC136279.1 | - | 2 | 326 | 9020266 | ||
BC142638.1 | - | 2 | 659 | |||
BC142688.1 | - | 2 | 608 | 40148654 | ||
CN350368.1 | - | 2 | 667 |