Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039380.3 | NP_001034469.2 | 1 | 343 | ||
NR_126421.1 | 1 | 343 | ||||
XM_017015895.1 | XP_016871384.1 | 1 | 337 | |||
GenBank mRNA | AK055129.1 | - | 1 | 321 | ||
BC130369.1 | - | 1 | 317 | 40146400 | ||
BC130395.1 | - | 1 | 317 | 40146426 | ||
DA772947.1 | - | 1 | 190 | |||
DR005042.1 | - | 1 | 461 | |||
HY023488.1 | - | 1 | 343 |