Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286574.1 | NP_001273503.1 | 2 | 300 | ||
NM_001286576.1 | NP_001273505.1 | 2 | 300 | |||
NM_145028.4 | NP_659465.2 | 2 | 381 | |||
XM_011514381.2 | XP_011512683.1 | 2 | 249 | |||
XM_011514382.2 | XP_011512684.1 | 2 | 380 | |||
XM_017010435.1 | XP_016865924.1 | 3 | 1855 | |||
GenBank mRNA | AK058119.1 | - | 2 | 250 | ||
BC033033.2 | - | 2 | 331 | 5268250 | ||
CR735810.1 | - | 2 | 194 | |||
DB070109.1 | - | 2 | 300 |