Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037325.2 | NP_001032402.1 | 4, 5 | 711 | ||
NM_001199165.2 | NP_001186094.1 | 24, 25 | 2842 | |||
NM_001302891.1 | NP_001289820.1 | 22, 23 | 2792 | |||
NR_126542.1 | 3, 4 | 541 | ||||
XM_005257119.4 | XP_005257176.1 | 24, 25 | 2936 | |||
XM_005257125.3 | XP_005257182.1 | 16, 17 | 1926 | |||
XM_005257126.4 | XP_005257183.1 | 13, 14 | 1591 | |||
XM_006721740.2 | XP_006721803.1 | 24, 25 | 2808 | |||
XM_011524461.2 | XP_011522763.1 | 24, 25 | 2949 | |||
XM_011524464.2 | XP_011522766.1 | 22, 23 | 2725 | |||
XM_011524465.2 | XP_011522767.1 | 21, 22 | 2265 | |||
XM_011524466.2 | XP_011522768.1 | 14, 15 | 1744 | |||
XM_011524467.2 | XP_011522769.1 | 13, 14 | 1586 | |||
XM_017024302.1 | XP_016879791.1 | 22, 23 | 2679 | |||
GenBank mRNA | AF458591.1 | - | 14, 15 | 1749 | ||
AK098070.1 | - | 14, 15 | 1661 | |||
BC029524.1 | - | 4, 5 | 711 | |||
BC131805.1 | - | 22, 23 | 2620 | 40119069 | ||
BG531260.1 | - | 1, 2 | 29 | |||
BX097629.1 | - | 4, 5 | 389 |