Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRA10AC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145246.4 | NP_660289.2 | 8, 9 | 711 | ||
XM_005269515.4 | XP_005269572.1 | 9, 10 | 808 | |||
XM_005269516.4 | XP_005269573.1 | 8, 9 | 903 | |||
XM_011539259.2 | XP_011537561.1 | 8, 9 | 676 | |||
XM_011539260.2 | XP_011537562.1 | 9, 10 | 766 | |||
XM_017015656.1 | XP_016871145.1 | 8, 9 | 676 | |||
XM_017015657.1 | XP_016871146.1 | 8, 9 | 711 | |||
XM_017015658.1 | XP_016871147.1 | 9, 10 | 811 | |||
XM_017015659.1 | XP_016871148.1 | 8, 9 | 903 | |||
XM_017015660.1 | XP_016871149.1 | 9, 10 | 768 | |||
XM_017015661.1 | XP_016871150.1 | 9, 10 | 825 | |||
XM_017015662.1 | XP_016871151.1 | 8, 9 | 676 | |||
GenBank mRNA | AJ431721.1 | - | 8, 9 | 707 | ||
AJ431722.1 | - | 8, 9 | 707 | |||
AJ431723.1 | - | 8, 9 | 707 | |||
AJ431724.1 | - | 8, 9 | 707 | |||
AJ431725.1 | - | 8, 9 | 707 | |||
AK090955.1 | - | 8, 9 | 711 | |||
BC018007.1 | - | 8, 9 | 580 | 4278022 |