Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144033.1 | NP_001137505.1 | 4 | 395 | ||
NM_145286.2 | NP_660329.1 | 3 | 311 | |||
GenBank mRNA | AK304159.1 | - | 4 | 395 | ||
AK313632.1 | - | 3 | 283 | |||
BC025760.1 | - | 3 | 298 | 5204252 | ||
DB200356.1 | - | 4 | 374 |