Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSMEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145268.3 | NP_660311.1 | 2 | 247 | ||
XM_005250141.3 | XP_005250198.1 | 2 | 154 | |||
XM_011515794.2 | XP_011514096.1 | 2 | 111 | |||
XM_011515795.2 | XP_011514097.1 | 2 | 104 | |||
GenBank mRNA | AK097635.1 | - | 2 | 233 | ||
BC017587.2 | - | 2 | 251 | 4838989 |