Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHLDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134437.1 | NP_001127909.1 | 6 | 2293 | ||
NM_001134438.1 | NP_001127910.1 | 5 | 2351 | |||
NM_001134439.1 | NP_001127911.1 | 5 | 2112 | |||
NM_145753.2 | NP_665696.1 | 5 | 2351 | |||
GenBank mRNA | AB209903.1 | - | 5 | 1993 | ||
AF506820.1 | - | 4 | 2055 | |||
AJ496194.1 | - | 4 | 1966 | |||
AK092996.1 | - | 5 | 2037 | |||
AK096151.1 | - | 5 | 2293 | |||
AK125555.1 | - | 4 | 720 | |||
AK296841.1 | - | 6 | 2293 | |||
AK315050.1 | - | 5 | 2262 | |||
BC069194.1 | - | 5 | 2240 | |||
BC142678.1 | - | 4 | 1958 | 40148225 | ||
BC150210.1 | - | 4 | 1958 | 40148898 | ||
CR749654.1 | - | 6 | 2276 |