Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRFAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203462.2 | NP_982287.1 | 1 | 369 | ||
XM_005247932.1 | XP_005247989.1 | 1 | 355 | |||
GenBank mRNA | AF155654.1 | - | 1 | 367 | ||
AF258591.1 | - | 1 | 317 | |||
AK312676.1 | - | 1 | 348 | |||
BC008087.1 | - | 1 | 316 | 3446352 | ||
KC858143.1 | - | 1 | 122 |