Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF3IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164281.2 | NP_001157753.1 | 8 | 2013 | ||
NM_001164283.2 | NP_001157755.1 | 3 | 295 | |||
NM_147200.2 | NP_671733.2 | 9 | 1804 | |||
NM_147686.3 | NP_679211.2 | 8 | 2016 | |||
XM_006715319.3 | XP_006715382.1 | 3 | 257 | |||
XM_011535386.1 | XP_011533688.1 | 9 | 2739 | |||
GenBank mRNA | AF136405.1 | - | 9 | 1712 | ||
AF136406.1 | - | 8 | 1803 | |||
AF136407.1 | - | 8 | 1803 | |||
AF272151.1 | - | 7 | 1536 | |||
AF274303.1 | - | 8 | 1536 | |||
AK025351.1 | - | 8 | 1610 | |||
AK026602.1 | - | 7 | 1010 | |||
AK314415.1 | - | 9 | 1727 | |||
AL050289.1 | - | 8 | 1688 | |||
BC002823.2 | - | 8 | 1599 | 3634505 | ||
BC026723.1 | - | 3 | 214 | |||
BI856094.1 | - | 3 | 293 | |||
BM449775.1 | - | 5 | 478 | |||
CB049654.1 | - | 3 | 262 |