Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134665.2 | NP_001128137.1 | 8 | 2705 | ||
NM_001134666.2 | NP_001128138.1 | 8 | 2694 | |||
NM_152292.4 | NP_689505.1 | 8 | 2850 | |||
XM_005263352.3 | XP_005263409.1 | 8 | 2684 | |||
XM_006714417.2 | XP_006714480.1 | 8 | 2726 | |||
GenBank mRNA | BC028373.2 | - | 8 | 2848 | 4838346 | |
EU668342.1 | - | 8 | 2848 |