Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152329.3 | NP_689542.2 | 4 | 1659 | ||
NM_203467.1 | NP_982292.1 | 3 | 937 | |||
NR_037792.1 | 6 | 1879 | ||||
NR_037793.1 | 5 | 1767 | ||||
GenBank mRNA | AK293156.1 | - | 4 | 1378 | ||
AY052405.1 | - | 4 | 1345 | |||
BC030142.2 | - | 3 | 873 | |||
BC064133.1 | - | 5 | 1512 | |||
BC107888.1 | - | 3 | 686 | |||
BC139921.1 | - | 4 | 1399 | 40148567 | ||
BQ059807.1 | - | 4 | 674 | |||
BX248298.1 | - | 4 | 1402 | |||
BX648029.1 | - | 6 | 1802 |