Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199144.1 | NP_001186073.1 | 7 | 8813 | ||
NM_152381.5 | NP_689594.4 | 9 | 9215 | |||
XM_017003309.1 | XP_016858798.1 | 7 | 8710 | |||
GenBank mRNA | AK056582.1 | - | 1 | 443 | ||
AK096907.1 | - | 1 | 1450 | |||
AK125495.1 | - | 1 | 2584 | |||
AL831970.2 | - | 1 | 1417 | |||
AL831985.1 | - | 1 | 1077 | |||
AL832011.1 | - | 1 | 1701 | |||
AL832336.1 | - | 1 | 1742 | |||
AL832360.1 | - | 1 | 1322 | |||
AL832375.1 | - | 1 | 753 | |||
AL832382.1 | - | 1 | 1269 | |||
AL832452.2 | - | 2 | 1790 | |||
AL833291.2 | - | 2 | 3033 | |||
AY820969.1 | - | 7 | 8682 | |||
BC070143.1 | - | 1 | 367 | |||
BC107799.1 | - | 1 | 286 | |||
BX647448.1 | - | 1 | 2765 | |||
BX647645.1 | - | 1 | 1436 | |||
BX647844.1 | - | 1 | 2616 | |||
BX647845.1 | - | 1 | 1374 | |||
BX647848.1 | - | 1 | 3254 | |||
EF119711.1 | - | 7 | 8813 |