Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152397.2 | NP_689610.2 | 4 | 616 | ||
XM_011533366.2 | XP_011531668.1 | 3 | 434 | |||
XM_017005729.1 | XP_016861218.1 | 3 | 445 | |||
GenBank mRNA | AK131018.1 | - | 4 | 567 | ||
BC029595.1 | - | 4 | 581 | 5273825 |