Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZG16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152338.3 | NP_689551.2 | 4 | 292 | ||
XM_011545921.2 | XP_011544223.1 | 4 | 371 | |||
GenBank mRNA | AB092813.1 | - | 3 | 218 | ||
AK125559.1 | - | 4 | 250 | |||
AK312002.1 | - | 3 | 240 | |||
BC029149.1 | - | 3 | 224 | 5187790 | ||
BC150656.1 | - | 4 | 263 | 9057027 | ||
CK904265.1 | - | 4 | 292 |