Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TACC2 SIRNA Assays ›
Gene Symbol:
TACC2
Gene Name:
transforming acidic coiled-coil containing protein 2
Gene Aliases:
AZU-1, ECTACC
Chromosome Location:
Chr.10: 121989174 - 122254545 on Build GRCh38
UniGene ID:
Hs.501252
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
3868
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TACC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291876.1
NP_001278805.1
20
8931
NM_001291877.1
NP_001278806.1
20
9078
NM_001291878.1
NP_001278807.1
17
3616
NM_001291879.1
NP_001278808.1
17
3289
NM_006997.3
NP_008928.1
17
3670
NM_206860.2
NP_996742.1
16
3580
NM_206861.2
NP_996743.1
20
3735
NM_206862.3
NP_996744.3
23
9297
XM_005269388.1
XP_005269445.1
24
9214
XM_005269389.4
XP_005269446.1
24
9738
XM_005269390.1
XP_005269447.1
25
9157
XM_005269391.1
XP_005269448.1
23
9154
XM_005269392.1
XP_005269449.1
23
9124
XM_005269393.1
XP_005269450.1
24
9121
XM_005269394.1
XP_005269451.1
23
9088
XM_005269395.1
XP_005269452.1
23
9079
XM_005269396.1
XP_005269453.1
22
8983
XM_005269397.1
XP_005269454.1
22
8953
XM_005269399.1
XP_005269456.1
21
8755
XM_006717548.1
XP_006717611.1
24
9109
XM_006717552.1
XP_006717615.1
23
8926
XM_006717553.1
XP_006717616.1
22
8890
XM_006717554.1
XP_006717617.1
22
8878
XM_011539126.1
XP_011537428.1
24
9211
XM_011539128.1
XP_011537430.1
22
3787
XM_011539129.1
XP_011537431.1
21
3652
XM_011539130.1
XP_011537432.1
21
3559
XM_011539131.1
XP_011537433.1
17
3625
XM_011539132.1
XP_011537434.1
18
3568
XM_011539133.1
XP_011537435.1
16
3242
XM_011539134.1
XP_011537436.1
16
3513
XM_017015484.1
XP_016870973.1
24
9202
XM_017015485.1
XP_016870974.1
24
9067
XM_017015486.1
XP_016870975.1
23
9031
XM_017015487.1
XP_016870976.1
23
9019
XM_017015488.1
XP_016870977.1
22
8848
XM_017015489.1
XP_016870978.1
21
8770
XM_017015491.1
XP_016870980.1
17
3765
XM_017015492.1
XP_016870981.1
17
3672
XM_017015493.1
XP_016870982.1
16
3430
XM_017015494.1
XP_016870983.1
15
3441
XM_017015495.1
XP_016870984.1
15
3301
XM_017015496.1
XP_016870985.1
18
3566
XM_017015497.1
XP_016870986.1
18
3825
XM_017015498.1
XP_016870987.1
17
3690
XM_017015499.1
XP_016870988.1
19
3659
GenBank mRNA
AF095791.2
-
17
3256
AF176646.1
-
16
3415
AF220152.2
-
13
3103
AF528098.1
-
20
3716
AF528099.1
-
23
9278
AK027066.1
-
13
2209
AK056743.1
-
4
1526
AK075097.1
-
13
1567
AK095371.1
-
4
2715
AL713712.1
-
13
2515
AL833304.1
-
22
8396
BC000999.2
-
6
664
BC039311.1
-
16
3580
BC141837.1
-
18
3268
BC142673.1
-
18
3268
BC144599.1
-
20
9046
9053129
BC144600.1
-
20
8899
9053130
BC150627.1
-
20
8899
9056997
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell proliferation
cerebral cortex development
nuclear hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0