Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKLE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278443.1 | NP_001265372.1 | 8 | 1730 | ||
NM_001278445.1 | NP_001265374.1 | 8 | 1758 | |||
NM_152363.5 | NP_689576.5 | 8 | 1841 | |||
NR_103530.1 | 7 | 1585 | ||||
GenBank mRNA | AK092706.1 | - | 8 | 1531 | ||
AK096688.1 | - | 4 | 810 | |||
AK303628.1 | - | 7 | 1585 | |||
BC125274.1 | - | 8 | 1498 | 40085198 | ||
EU184013.1 | - | 8 | 1565 |