Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152372.3 | NP_689585.3 | 30 | 3717 | ||
GenBank mRNA | AK093280.1 | - | 3 | 603 | ||
AK128233.1 | - | 22 | 2777 | |||
BC014671.1 | - | 4 | 237 | |||
BX537911.1 | - | 29 | 3441 | |||
BX647453.1 | - | 30 | 3678 | |||
BX647455.1 | - | 30 | 3717 | |||
BX647644.1 | - | 30 | 3678 |