Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152485.3 | NP_689698.1 | 2 | 888 | ||
GenBank mRNA | AB101206.1 | - | 2 | 888 | ||
AK057807.1 | - | 2 | 717 | |||
BC039719.1 | - | 2 | 703 | 5770046 |