Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152543.2 | NP_689756.2 | 2 | 266 | ||
XM_011531671.2 | XP_011529973.1 | 3 | 301 | |||
XM_011531672.2 | XP_011529974.1 | 3 | 411 | |||
XM_017007808.1 | XP_016863297.1 | 6 | 1167 | |||
XM_017007809.1 | XP_016863298.1 | 6 | 1097 | |||
XM_017007810.1 | XP_016863299.1 | 6 | 1273 | |||
GenBank mRNA | AK058100.1 | - | 2 | 266 | ||
BP369394.1 | - | 2 | 266 |