Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152482.2 | NP_689695.2 | 3 | 1206 | ||
XM_005259506.3 | XP_005259563.1 | 3 | 1222 | |||
XM_006722653.3 | XP_006722716.1 | 3 | 1181 | |||
XM_017026374.1 | XP_016881863.1 | 3 | 1529 | |||
GenBank mRNA | AK056004.1 | - | 1 | 2234 | ||
AK075267.1 | - | 3 | 1189 | |||
AK093985.1 | - | 4 | 1083 | |||
AK297944.1 | - | 3 | 1314 | |||
AL122089.1 | - | 3 | 1177 | |||
AY007120.1 | - | 2 | 1142 | |||
BC018441.1 | - | 3 | 1176 | 4471390 |