Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152556.2 | NP_689769.2 | 2 | 408 | ||
GenBank mRNA | AK056380.1 | - | 2 | 408 | ||
AK299901.1 | - | 4 | 254 | |||
AL834437.1 | - | 2 | 203 | |||
BC114615.1 | - | 2 | 400 | 40032372 | ||
DA738444.1 | - | 2 | 408 |