Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258282.1 | NP_001245211.1 | 7 | 918 | ||
NM_152570.2 | NP_689783.1 | 7 | 998 | |||
XM_011517719.1 | XP_011516021.1 | 6 | 1866 | |||
XM_011517724.2 | XP_011516026.1 | 5 | 4885 | |||
XM_011517728.2 | XP_011516030.1 | 2 | 4664 | |||
XM_017014303.1 | XP_016869792.1 | 11 | 4769 | |||
XM_017014304.1 | XP_016869793.1 | 4 | 7289 | |||
XM_017014305.1 | XP_016869794.1 | 5 | 6872 | |||
XM_017014306.1 | XP_016869795.1 | 3 | 4033 | |||
XM_017014307.1 | XP_016869796.1 | 2 | 1506 | |||
GenBank mRNA | AK056372.1 | - | 7 | 998 | ||
AK290972.1 | - | 2 | 529 | |||
AY358114.1 | - | 2 | 552 | |||
BC137514.1 | - | 7 | 858 | 9021514 | ||
BC144676.1 | - | 7 | 858 | 9053210 |