Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152616.4 | NP_689829.3 | 2 | 748 | ||
XM_011512740.2 | XP_011511042.1 | 2 | 743 | |||
GenBank mRNA | AF521868.1 | - | 2 | 560 | ||
AK097416.1 | - | 2 | 735 | |||
AK310536.1 | - | 2 | 727 | |||
AL833848.1 | - | 1 | 34 | |||
BC130472.1 | - | 2 | 658 | 40146503 | ||
BC130474.1 | - | 2 | 658 | 40146505 |