Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297598.1 | NP_001284527.1 | 5 | 394 | ||
NM_001297599.1 | NP_001284528.1 | 5 | 379 | |||
NM_152622.4 | NP_689835.3 | 5 | 379 | |||
XM_011543216.1 | XP_011541518.1 | 5 | 414 | |||
XM_011543217.1 | XP_011541519.1 | 5 | 414 | |||
XM_017009149.1 | XP_016864638.1 | 6 | 415 | |||
GenBank mRNA | AK093273.1 | - | 3 | 306 | ||
AK299285.1 | - | 4 | 354 | |||
BC041348.2 | - | 5 | 376 | 5272287 | ||
BX537798.1 | - | 5 | 364 | |||
BX640907.1 | - | 5 | 382 | |||
BX647359.1 | - | 5 | 373 | |||
BX648294.1 | - | 5 | 374 | |||
CR749246.1 | - | 6 | 414 | |||
CR749805.1 | - | 6 | 414 | |||
DB136129.1 | - | 5 | 383 |