Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178007.1 | NP_001171478.1 | 3 | 1875 | ||
NM_152618.2 | NP_689831.2 | 2 | 1687 | |||
XM_011531680.2 | XP_011529982.1 | 2 | 1634 | |||
XM_017007831.1 | XP_016863320.1 | 2 | 1702 | |||
GenBank mRNA | AK092949.1 | - | 1 | 801 | ||
AK123553.1 | - | 2 | 1687 | |||
BC055426.1 | - | 2 | 1685 | 5262398 | ||
BX538148.1 | - | 2 | 1626 |