Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIKFYVE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178000.1 | NP_001171471.1 | 12 | 1802 | ||
NM_152671.3 | NP_689884.1 | 10 | 1511 | |||
GenBank mRNA | BC032389.1 | - | 10 | 1511 | ||
BC125052.1 | - | 11 | 1533 | |||
BC125053.1 | - | 12 | 1791 |