Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC51A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152672.5 | NP_689885.4 | 5, 6 | 730 | ||
GenBank mRNA | AK172837.1 | - | 5, 6 | 733 | ||
AK310276.1 | - | 2, 3 | 381 | |||
AY194243.1 | - | 5, 6 | 551 | |||
BC029606.1 | - | 1, 2 | 342 | 5297443 |