Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152751.2 | NP_689964.2 | 9 | 1557 | ||
XM_005252402.4 | XP_005252459.1 | 9 | 1655 | |||
XM_011519390.2 | XP_011517692.1 | 8 | 1348 | |||
XM_011519391.2 | XP_011517693.1 | 8 | 1345 | |||
XM_011519392.2 | XP_011517694.1 | 9 | 1651 | |||
XM_011519393.2 | XP_011517695.1 | 8 | 1443 | |||
XM_011519394.2 | XP_011517696.1 | 10 | 1590 | |||
XM_011519395.2 | XP_011517697.1 | 8 | 1950 | |||
XM_011519396.2 | XP_011517698.1 | 9 | 2041 | |||
XM_011519397.2 | XP_011517699.1 | 9 | 1596 | |||
XM_017015915.1 | XP_016871404.1 | 10 | 1575 | |||
GenBank mRNA | AK097602.1 | - | 9 | 1557 |