Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIBADH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152740.3 | NP_689953.1 | 5 | 750 | ||
XM_017011719.1 | XP_016867208.1 | 6 | 780 | |||
GenBank mRNA | AB050000.1 | - | 5 | 624 | ||
AF529362.1 | - | 5 | 537 | |||
AK025558.1 | - | 4 | 362 | |||
AK316605.1 | - | 5 | 555 | |||
BC013855.1 | - | 2 | 123 | |||
BC020167.1 | - | 5 | 611 | |||
BC032324.1 | - | 5 | 635 | 3890576 |