Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN7L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152749.3 | NP_689962.1 | 4 | 1342 | ||
XM_011515954.2 | XP_011514256.1 | 5 | 1425 | |||
GenBank mRNA | AK027276.1 | - | 1 | 460 | ||
AK095475.1 | - | 4 | 1257 | |||
BC030616.2 | - | 4 | 1339 | 4796569 | ||
BP339600.1 | - | 1 | 134 |