Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CADM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167674.1 | NP_001161146.1 | 2 | 704 | ||
NM_001167675.1 | NP_001161147.1 | 3 | 731 | |||
NM_001256502.1 | NP_001243431.1 | 3 | 251 | |||
NM_001256504.1 | NP_001243433.1 | 4 | 278 | |||
NM_001256505.1 | NP_001243434.1 | 3 | 251 | |||
NM_153184.3 | NP_694854.2 | 2 | 84 | |||
XM_006713081.3 | XP_006713144.1 | 3 | 594 | |||
XM_006713082.3 | XP_006713145.1 | 2 | 697 | |||
XM_017006062.1 | XP_016861551.1 | 4 | 636 | |||
XM_017006063.1 | XP_016861552.1 | 2 | 284 | |||
GenBank mRNA | AB646739.1 | - | 3 | 731 | ||
AB646740.1 | - | 3 | 251 | |||
AB646741.1 | - | 3 | 251 | |||
AB646742.1 | - | 4 | 278 | |||
AF538973.1 | - | 2 | 84 | |||
AK295885.1 | - | 3 | 727 | |||
AL834270.1 | - | 2 | 674 | |||
BC105999.1 | - | 3 | 731 | 5259422 | ||
BC112136.1 | - | 2 | 140 | 8327604 | ||
BC112138.1 | - | 2 | 152 | 8327606 |