Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSMEM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304385.1 | NP_001291314.1 | 4 | 503 | ||
NM_153215.2 | NP_694947.1 | 4 | 506 | |||
XM_006712979.3 | XP_006713042.1 | 4 | 982 | |||
XM_006712980.3 | XP_006713043.1 | 4 | 979 | |||
XM_011533370.2 | XP_011531672.1 | 4 | 758 | |||
GenBank mRNA | AK095927.1 | - | 4 | 433 | ||
BC028000.1 | - | 4 | 446 | 5197901 | ||
DA830939.1 | - | 4 | 503 |