Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TANGO2 SIRNA Assays ›
Gene Symbol:
TANGO2
Gene Name:
transport and golgi organization 2 homolog
Gene Aliases:
C22orf25, MECRCN
Chromosome Location:
Chr.22: 20017000 - 20067164 on Build GRCh38
UniGene ID:
Hs.474233
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
39829
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TANGO2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001283106.2
NP_001270035.1
5
522
NM_001283116.2
NP_001270045.1
4
276
NM_001283129.2
NP_001270058.1
4
516
NM_001283148.2
NP_001270077.1
4
276
NM_001283154.2
NP_001270083.1
4
388
NM_001283179.2
NP_001270108.1
4
403
NM_001283186.2
NP_001270115.1
4
276
NM_001283199.2
NP_001270128.1
4
388
NM_001283215.2
NP_001270144.1
4
516
NM_001283235.2
NP_001270164.1
4
276
NM_001283248.2
NP_001270177.1
4
276
NM_001322141.1
NP_001309070.1
4
516
NM_001322142.1
NP_001309071.1
4
276
NM_001322143.1
NP_001309072.1
4
589
NM_001322144.1
NP_001309073.1
4
516
NM_001322145.1
NP_001309074.1
4
589
NM_001322146.1
NP_001309075.1
4
349
NM_001322147.1
NP_001309076.1
4
516
NM_001322148.1
NP_001309077.1
4
516
NM_001322149.1
NP_001309078.1
4
516
NM_001322150.1
NP_001309079.1
4
516
NM_001322153.1
NP_001309082.1
4
276
NM_001322155.1
NP_001309084.1
4
349
NM_001322160.1
NP_001309089.1
4
401
NM_001322163.1
NP_001309092.1
5
522
NM_001322166.1
NP_001309095.1
4
349
NM_001322167.1
NP_001309096.1
4
401
NM_001322169.1
NP_001309098.1
4
276
NM_001322171.1
NP_001309100.1
4
349
NM_001322172.1
NP_001309101.1
4
641
NM_001322173.1
NP_001309102.1
4
516
NM_001322174.1
NP_001309103.1
4
403
NM_001322175.1
NP_001309104.1
4
401
NM_152906.6
NP_690870.3
4
388
NR_104274.2
4
276
NR_104275.2
4
516
NR_136206.1
4
349
NR_136211.1
4
349
NR_136212.1
4
349
XM_011529863.1
XP_011528165.1
4
350
XM_011529865.1
XP_011528167.1
3
292
XM_011529867.1
XP_011528169.1
5
486
XM_017028577.1
XP_016884066.1
4
349
XM_017028578.1
XP_016884067.1
4
368
XM_017028579.1
XP_016884068.1
4
397
XM_017028580.1
XP_016884069.1
5
486
XM_017028581.1
XP_016884070.1
4
403
XM_017028582.1
XP_016884071.1
5
486
XM_017028583.1
XP_016884072.1
4
233
XM_017028584.1
XP_016884073.1
4
496
XM_017028585.1
XP_016884074.1
4
348
XM_017028586.1
XP_016884075.1
4
273
XM_017028587.1
XP_016884076.1
4
403
XM_017028588.1
XP_016884077.1
4
329
GenBank mRNA
AK092484.1
-
4
231
AK289862.1
-
5
491
AK295210.1
-
3
402
AK297075.1
-
4
233
AK297922.1
-
4
234
AK298593.1
-
4
234
AK301054.1
-
4
235
AK301366.1
-
4
475
AK309398.1
-
3
156
AK311676.1
-
4
235
AK316020.1
-
4
237
AK316056.1
-
4
496
AL713640.1
-
1
16
BC041339.1
-
4
351
5284126
BG685068.1
-
2
126
BI544307.1
-
4
351
BI911898.1
-
4
339
BM013784.1
-
4
560
BP193987.1
-
4
474
BP303570.1
-
3
173
BU173566.1
-
4
308
BX345246.2
-
4
388
CD675132.1
-
4
330
CF552364.1
-
3
247
CR456355.1
-
3
203
CR627452.1
-
4
231
DA176876.1
-
4
274
DB021425.1
-
2
66
DC301369.1
-
4
245
DC303300.1
-
5
491
DC315148.1
-
4
234
DC318353.1
-
4
276
DC332363.1
-
4
496
DC349456.1
-
4
472
HY135099.1
-
4
403
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0