Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166271.2 | NP_001159743.1 | 5 | 2556 | ||
NM_001286792.1 | NP_001273721.1 | 7 | 2880 | |||
NM_001286793.1 | NP_001273722.1 | 2 | 290 | |||
NM_001286794.1 | NP_001273723.1 | 3 | 498 | |||
NM_001286795.1 | NP_001273724.1 | 2 | 290 | |||
NM_153023.3 | NP_694568.1 | 4 | 792 | |||
GenBank mRNA | AK055770.1 | - | 4 | 760 | ||
AK123031.1 | - | 2 | 190 | |||
AK297394.1 | - | 2 | 260 | |||
AK298717.1 | - | 3 | 400 | |||
AK299981.1 | - | 2 | 233 | |||
AK301149.1 | - | 2 | 233 | |||
BC109290.1 | - | 3 | 502 | 40034357 | ||
BC109291.1 | - | 3 | 502 | 40034359 | ||
BX648244.1 | - | 7 | 2702 |