Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000044.3 | NP_000035.2 | 1 | 2453 | ||
GenBank mRNA | AF162704.1 | - | 2 | 1320 | ||
AF321914.1 | - | 1 | 1428 | |||
AF321915.1 | - | 1 | 1416 | |||
AF321916.1 | - | 1 | 1425 | |||
AF321917.1 | - | 2 | 1392 | |||
BC132975.1 | - | 1 | 1795 | 40146997 | ||
FJ235916.1 | - | 1 | 1680 | |||
FJ235917.1 | - | 1 | 1677 | |||
FJ235918.1 | - | 1 | 1677 | |||
FJ235919.1 | - | 1 | 1680 | |||
FJ235920.1 | - | 1 | 1680 | |||
GU208210.1 | - | 1 | 1344 | |||
HM055487.1 | - | 1 | 1680 | |||
KJ534771.1 | - | 1 | 1335 | |||
L29496.1 | - | 1 | 768 | |||
M20132.1 | - | 1 | 1694 | |||
M21748.1 | - | 1 | 1491 | |||
M23263.1 | - | 1 | 1851 | |||
M34233.1 | - | 2 | 1425 | |||
M73069.1 | - | 1 | 1383 |