Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080571.1 | NP_542138.1 | 3 | 279 | ||
NM_153269.2 | NP_695001.2 | 3 | 275 | |||
XM_006723543.3 | XP_006723606.1 | 2 | 205 | |||
GenBank mRNA | AK091924.1 | - | 2 | 204 | ||
AK097394.1 | - | 3 | 246 | |||
BC051762.1 | - | 2 | 224 | |||
BC134417.1 | - | 3 | 233 | 40115967 | ||
BC137450.1 | - | 3 | 242 | 9021450 | ||
BC144648.1 | - | 2 | 193 | |||
BC144649.1 | - | 3 | 246 | 9053180 | ||
BX640715.1 | - | 2 | 219 | |||
DB226405.1 | - | 3 | 275 |