Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286233.1 | NP_001273162.1 | 16 | 2603 | ||
NM_001286234.1 | NP_001273163.1 | 11 | 1849 | |||
NM_001286235.1 | NP_001273164.1 | 13 | 2147 | |||
NM_001286236.1 | NP_001273165.1 | 10 | 1688 | |||
NM_001286237.1 | NP_001273166.1 | 10 | 1742 | |||
NM_153449.3 | NP_703150.1 | 12 | 1885 | |||
XM_005253315.3 | XP_005253372.1 | 11 | 1683 | |||
XM_005253317.4 | XP_005253374.1 | 11 | 2202 | |||
XM_011520562.1 | XP_011518864.1 | 14 | 2037 | |||
XM_011520563.2 | XP_011518865.1 | 11 | 1697 | |||
XM_011520564.2 | XP_011518866.1 | 10 | 1506 | |||
XM_011520565.2 | XP_011518867.1 | 9 | 1535 | |||
XM_017018841.1 | XP_016874330.1 | 12 | 1782 | |||
XM_017018842.1 | XP_016874331.1 | 15 | 2353 | |||
XM_017018843.1 | XP_016874332.1 | 15 | 2677 | |||
XM_017018844.1 | XP_016874333.1 | 12 | 1732 | |||
XM_017018845.1 | XP_016874334.1 | 12 | 2274 | |||
XM_017018846.1 | XP_016874335.1 | 12 | 1858 | |||
XM_017018847.1 | XP_016874336.1 | 11 | 1815 | |||
GenBank mRNA | AF481878.1 | - | 11 | 1668 | ||
AF481879.1 | - | 12 | 1745 | |||
AK126026.1 | - | 16 | 2385 | |||
AK302881.1 | - | 10 | 1501 | |||
AL110298.1 | - | 5 | 928 | |||
AY357941.1 | - | 11 | 1834 | |||
BC060766.1 | - | 11 | 1696 | 5297510 |