Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBSPON | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153225.3 | NP_694957.3 | 3, 4 | 705 | ||
XM_017013145.1 | XP_016868634.1 | 3, 4 | 950 | |||
GenBank mRNA | AK292970.1 | - | 3, 4 | 605 | ||
AY040546.1 | - | 3, 4 | 603 | |||
BC042877.1 | - | 3, 4 | 705 | 5169941 |