Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122890.2 | NP_001116362.1 | 4 | 771 | ||
NM_001288702.1 | NP_001275631.1 | 4 | 789 | |||
NM_153338.3 | NP_699169.2 | 3 | 675 | |||
GenBank mRNA | AK074646.1 | - | 3 | 660 | ||
AK296645.1 | - | 4 | 770 | |||
AK298329.1 | - | 3 | 752 | |||
AK300648.1 | - | 4 | 749 | |||
BC063111.1 | - | 4 | 741 | 4593403 |