Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM86A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153347.2 | NP_699178.1 | 2, 3 | 431 | ||
GenBank mRNA | AK074600.1 | - | 2, 3 | 387 | ||
BC017073.2 | - | 1, 2 | 219 | |||
BC035692.1 | - | 2, 3 | 362 | 5759114 | ||
DA047874.1 | - | 2, 3 | 431 |