Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286233.1 | NP_001273162.1 | 16 | 2630 | ||
NM_001286234.1 | NP_001273163.1 | 11 | 1876 | |||
NM_001286235.1 | NP_001273164.1 | 13 | 2174 | |||
NM_001286236.1 | NP_001273165.1 | 10 | 1715 | |||
NM_001286237.1 | NP_001273166.1 | 10 | 1769 | |||
NM_153449.3 | NP_703150.1 | 12 | 1912 | |||
XM_005253315.3 | XP_005253372.1 | 11 | 1710 | |||
XM_005253317.4 | XP_005253374.1 | 11 | 2229 | |||
XM_011520562.1 | XP_011518864.1 | 14 | 2064 | |||
XM_011520563.2 | XP_011518865.1 | 11 | 1724 | |||
XM_011520564.2 | XP_011518866.1 | 10 | 1533 | |||
XM_011520565.2 | XP_011518867.1 | 9 | 1562 | |||
XM_017018841.1 | XP_016874330.1 | 12 | 1809 | |||
XM_017018842.1 | XP_016874331.1 | 15 | 2380 | |||
XM_017018843.1 | XP_016874332.1 | 15 | 2704 | |||
XM_017018844.1 | XP_016874333.1 | 12 | 1759 | |||
XM_017018845.1 | XP_016874334.1 | 12 | 2301 | |||
XM_017018846.1 | XP_016874335.1 | 12 | 1885 | |||
XM_017018847.1 | XP_016874336.1 | 11 | 1842 | |||
GenBank mRNA | AF481878.1 | - | 11 | 1695 | ||
AF481879.1 | - | 12 | 1772 | |||
AL110298.1 | - | 5 | 955 | |||
BC060766.1 | - | 11 | 1723 | 5297510 |