Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317078.1 | NP_001304007.1 | 5 | 1114 | ||
NM_153450.2 | NP_703151.2 | 4 | 1548 | |||
GenBank mRNA | AY148462.1 | - | 5 | 843 | ||
BC037223.1 | - | 4 | 1426 | 5197938 |