Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF3R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000760.3 | NP_000751.1 | 7 | 1367 | ||
NM_156039.3 | NP_724781.1 | 7 | 1367 | |||
NM_172313.2 | NP_758519.1 | 7 | 1367 | |||
XM_005270493.1 | XP_005270550.1 | 7 | 1367 | |||
XM_011540748.2 | XP_011539050.1 | 6 | 892 | |||
XM_011540749.1 | XP_011539051.1 | 7 | 1367 | |||
XM_011540750.1 | XP_011539052.1 | 2 | 197 | |||
XM_017000370.1 | XP_016855859.1 | 6 | 929 | |||
GenBank mRNA | AB209656.1 | - | 3 | 2166 | ||
BC053585.1 | - | 7 | 960 | 5217305 | ||
BX370109.2 | - | 2 | 188 | |||
DQ424902.1 | - | 5 | 783 | |||
M59818.1 | - | 7 | 952 | |||
M59819.1 | - | 7 | 952 | |||
M59820.1 | - | 7 | 952 | |||
X55720.1 | - | 7 | 947 | |||
X55721.1 | - | 7 | 948 |