Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153487.3 | NP_705691.1 | 17 | 7143 | ||
XM_017010734.1 | XP_016866223.1 | 17 | 6105 | |||
GenBank mRNA | AF478693.1 | - | 17 | 7143 | ||
AK055376.1 | - | 1 | 915 | |||
AK091650.1 | - | 3 | 900 | |||
AK092308.1 | - | 1 | 1587 | |||
AK126965.1 | - | 3 | 4585 | |||
BX647180.1 | - | 5 | 3723 | |||
BX647253.1 | - | 5 | 3723 |